1,4-Dimethoxy-2-butyne ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D4951-1ML D4951-5ML
TCIAD4951-1MLEA 58.3 EUR
TCIAD4951-1ML TCIAD4951-5ML
1,4-Dimethoxy-2-butyne ≥98.0% (by GC)
1,4-Dimethoxy-2-butyne
Formule: C₆H₁₀O₂
Kookpunt: 157°C
Smeltpunt: –39°C
Vlampunt: 53°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00048069
CAS nummer: 16356-02-8
UN: 3271
ADR: 3,III
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %