2,4-Dibromobenzyl alcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (2,4-Dibromophenyl)methanol

D5002-1G D5002-5G
TCIAD5002-1GEA 46.2 EUR
TCIAD5002-1G TCIAD5002-5G
2,4-Dibromobenzyl alcohol ≥98.0% (by GC)
2,4-Dibromobenzyl alcohol
Formule: C₇H₆Br₂O
MDL Nummer: MFCD09996906
CAS nummer: 666747-06-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C
Solubility in N,N-DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR