Dimethyl phenylphosphonite ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Phenyldimethoxyphosphine

D5014-1G D5014-5G
TCIAD5014-1GEA 20.9 EUR
TCIAD5014-1G TCIAD5014-5G
Dimethyl phenylphosphonite ≥98.0% (by GC)
Dimethyl phenylphosphonite
Formule: C₈H₁₁O₂P
MW: 170.15 g/mol
Kookpunt: 102°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008352
CAS nummer: 2946-61-4
EINECS: 220-960-6
UN: 1760
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67fb078402c7372fa701bd7502102fbea3e5b6a4&vid=67fb078402c7372fa701bd7502102fbea3e5b6a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR