9,10-Dihydro-9,9-dimethylacridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D5028-200MG D5028-1G D5028-5G
TCIAD5028-200MGEA 40.7 EUR
TCIAD5028-200MG TCIAD5028-1G TCIAD5028-5G
9,10-Dihydro-9,9-dimethylacridine ≥98.0% (by GC)
9,10-Dihydro-9,9-dimethylacridine
Formule: C₁₅H₁₅N
Kookpunt: 155°C
MDL Nummer: MFCD00030130
CAS nummer: 6267-02-3
EINECS: 228-436-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2008cbd94ff869cfb847d8acfb636c79003c5b12&vid=2008cbd94ff869cfb847d8acfb636c79003c5b12&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C