1,5-Dimethyl-2,4-bis[2-(trimethylsilyl)ethynyl]benzene ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D5029-1G D5029-5G
TCIAD5029-1GEA 55 EUR
TCIAD5029-1G TCIAD5029-5G
1,5-Dimethyl-2,4-bis[2-(trimethylsilyl)ethynyl]benzene ≥97.0% (by GC)
1,5-Dimethyl-2,4-bis[2-(trimethylsilyl)ethynyl]benzene
Formule: C₁₈H₂₆Si₂
Kookpunt: 140°C
CAS nummer: 1379822-08-8
Order Now
Specification Test Results
Purity (GC) min. 97.0 %
Melting point 55.0 to 59.0 deg-C