2,2-Difluoroethyl acetate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D5040-5G D5040-25G
TCIAD5040-5GEA 164 EUR
TCIAD5040-5G TCIAD5040-25G
2,2-Difluoroethyl acetate ≥98.0% (by GC)
2,2-Difluoroethyl acetate
Formule: C₄H₆F₂O₂
Kookpunt: 106°C
MDL Nummer: MFCD00069133
CAS nummer: 1550-44-3
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a0d35edaf392cc2f759d829aa7f33c8fd1402ae&vid=6a0d35edaf392cc2f759d829aa7f33c8fd1402ae&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %