Dexmedetomidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Dex, (S)-Medetomidine hydrochloride, (+)-Medetomidine hydrochloride, (S)-4-[1-(2,3-Dimethylphenyl)ethyl]-1H-imidazole hydrochloride, (S)-4-(α,2,3-Trimethybenzyl)-1H-imidazole hydrochloride, Precedex, 5-[(1S)-1-(2,3-Dimethylphenyl)ethyl]-1H-imidazole hydrochloride

D5062-20MG D5062-100MG
TCIAD5062-20MGEA 39.6 EUR
TCIAD5062-20MG TCIAD5062-100MG
Dexmedetomidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Dexmedetomidine hydrochloride
Formule: C₁₃H₁₆N₂·HCl
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD12912677
CAS nummer: 145108-58-3
Merck Index: 14,02946

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 156.0 to 160.0 deg-C
Specific rotation [a]20/D +49.0 to +54.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR