N,N-Diethyl-2,2,2-trifluoroacetamide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D5063-5G D5063-25G
TCIAD5063-5GEA 67.1 EUR
TCIAD5063-5G TCIAD5063-25G
N,N-Diethyl-2,2,2-trifluoroacetamide ≥98.0% (by GC)
N,N-Diethyl-2,2,2-trifluoroacetamide
Formule: C₆H₁₀F₃NO
MW: 169.15 g/mol
Kookpunt: 160°C
Vlampunt: 43°C
MDL Nummer: MFCD00192034
CAS nummer: 360-92-9
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR