4-[(6,7-Dimethoxyquinolin-4-yl)oxy]aniline

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-(4-Aminophenoxy)-6,7-dimethoxyquinoline, 4-((6,7-Dimethoxyquinolin-4-yl)oxy)aniline

D5070-200MG D5070-1G
TCIAD5070-200MGEA 89.1 EUR
TCIAD5070-200MG TCIAD5070-1G
4-[(6,7-Dimethoxyquinolin-4-yl)oxy]aniline
4-[(6,7-Dimethoxyquinolin-4-yl)oxy]aniline
Formule: C₁₇H₁₆N₂O₃
Smeltpunt: 214°C
MDL Nummer: MFCD19685633
CAS nummer: 190728-25-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f660a16fac436b8e40c05cc28102d880e4b9088&vid=9f660a16fac436b8e40c05cc28102d880e4b9088&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 211.0 to 215.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR