3',5'-Dichloro-2,2,2-trifluoroacetophenone ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1-(3,5-Dichlorophenyl)-2,2,2-trifluoroethanone

D5113-1G D5113-5G
TCIAD5113-1GEA 77 EUR
TCIAD5113-1G TCIAD5113-5G
3',5'-Dichloro-2,2,2-trifluoroacetophenone ≥96.0% (by GC)
3',5'-Dichloro-2,2,2-trifluoroacetophenone
Formule: C₈H₃Cl₂F₃O
Kookpunt: 87°C
MDL Nummer: MFCD01319996
CAS nummer: 130336-16-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b147b85987162a67c58e268101ce98d5501c0607&vid=b147b85987162a67c58e268101ce98d5501c0607&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR