(S)-2,4-Dichloro-alpha-(chloromethyl)benzyl alcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (S)-alpha-(Chloromethyl)-2,4-dichlorobenzyl alcohol, (S)-2-Chloro-1-(2,4-dichlorophenyl)ethanol

D5115-1G D5115-200MG
TCIAD5115-1GEA 184 EUR
TCIAD5115-1G TCIAD5115-200MG
(S)-2,4-Dichloro-alpha-(chloromethyl)benzyl alcohol ≥98.0% (by GC)
(S)-2,4-Dichloro-alpha-(chloromethyl)benzyl alcohol
Formule: C₈H₇Cl₃O
MW: 225.49 g/mol
MDL Nummer: MFCD07776655
CAS nummer: 126534-31-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C
Specific rotation [a]20/D +55.0 to +61.0 deg (C=1.2, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR