1,9-Dichlorononane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D5116-5G D5116-25G
TCIAD5116-5GEA 31.9 EUR
TCIAD5116-5G TCIAD5116-25G
1,9-Dichlorononane ≥98.0% (by GC)
1,9-Dichlorononane
Formule: C₉H₁₈Cl₂
Kookpunt: 150°C
MDL Nummer: MFCD00001024
CAS nummer: 821-99-8
EINECS: 212-484-2
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66529fcccbc97150fecfea06f472157edd6ed64e&vid=66529fcccbc97150fecfea06f472157edd6ed64e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR