1,3-Difluoro-5-propylbenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D5134-1G D5134-5G
TCIAD5134-1GEA 53.9 EUR
TCIAD5134-1G TCIAD5134-5G
1,3-Difluoro-5-propylbenzene ≥98.0% (by GC)
1,3-Difluoro-5-propylbenzene
Formule: C₉H₁₀F₂
Kookpunt: 170°C
MDL Nummer: MFCD12405568
CAS nummer: 183245-00-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94eb32984c6a0830a7b8c6b12d7977e693b048a7&vid=94eb32984c6a0830a7b8c6b12d7977e693b048a7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %