3,8-Dimethylacenaphthenequinone ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D5156-200MG D5156-1G
TCIAD5156-200MGEA 90.2 EUR
TCIAD5156-200MG TCIAD5156-1G
3,8-Dimethylacenaphthenequinone ≥97.0% (by GC)
3,8-Dimethylacenaphthenequinone
Formule: C₁₄H₁₀O₂
CAS nummer: 146885-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f26c91a3b8e942048591d114fdefbf1a250eb5b&vid=5f26c91a3b8e942048591d114fdefbf1a250eb5b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 205.0 to 209.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR