2,3-Dimethyl-4-fluoronitrobenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D5187-1G D5187-5G
TCIAD5187-1GEA 42.9 EUR
TCIAD5187-1G TCIAD5187-5G
2,3-Dimethyl-4-fluoronitrobenzene ≥98.0% (by GC)
2,3-Dimethyl-4-fluoronitrobenzene
Formule: C₈H₈FNO₂
Kookpunt: 110°C
MDL Nummer: MFCD05663712
CAS nummer: 1736-87-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79a6dc85b164cb321a6b06c6762217cd602d4476&vid=79a6dc85b164cb321a6b06c6762217cd602d4476&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C