3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D5239-1G
D5239-5G
TCIAD5239-1GEA
75.9
EUR
TCIAD5239-1G
TCIAD5239-5G
3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester
Formule:
C₁₆H₂₁BO₆ |
MDL Nummer:
MFCD11858596 CAS nummer: 944392-68-1 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...