2,5-Dibromo-3-tetradecylthiophene ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D5241-1G
D5241-5G
TCIAD5241-1GEA
90.2
EUR
TCIAD5241-1G
TCIAD5241-5G
2,5-Dibromo-3-tetradecylthiophene ≥97.0% (by GC)
2,5-Dibromo-3-tetradecylthiophene
Formule:
C₁₈H₃₀Br₂S |
CAS nummer:
160096-74-2 |
|
Specification Test Results
Appearance | White to Yellow to Orange powder to lump |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...