1,7-Dichloro-1,1,3,3,5,5,7,7-octamethyltetrasiloxane ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D5267-5G D5267-25G
TCIAD5267-5GEA 204 EUR
TCIAD5267-5G TCIAD5267-25G
1,7-Dichloro-1,1,3,3,5,5,7,7-octamethyltetrasiloxane ≥97.0% (by GC)
1,7-Dichloro-1,1,3,3,5,5,7,7-octamethyltetrasiloxane
Formule: C₈H₂₄Cl₂O₃Si₄
Kookpunt: 222°C
Smeltpunt: –62°C
Vlampunt: 87°C
MDL Nummer: MFCD00045148
CAS nummer: 2474-02-4
EINECS: 219-597-6
UN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=566e3e7d0d12f7b13065e2b4002974847871f193&vid=566e3e7d0d12f7b13065e2b4002974847871f193&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR