2-Decyl-1-tetradecanol ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Decyltetradecan-1-ol
D5283-25ML D5283-500ML
TCIAD5283-25MLEA 29.7 EUR
TCIAD5283-25ML TCIAD5283-500ML
2-Decyl-1-tetradecanol ≥97.0% (by GC)
2-Decyl-1-tetradecanol
Formule: C₂₄H₅₀O
MW: 354.66 g/mol
Kookpunt: 177°C
Vlampunt: 209°C
MDL Nummer: MFCD00674096
CAS nummer: 58670-89-6
EINECS: 261-385-0
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %