1,1,3-Trimethoxypropane ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B23448.06 B23448.14
B23448.06EA 39 EUR
B23448.06 B23448.14
1,1,3-Trimethoxypropane ≥98%
1,1,3-Trimethoxypropane

Purity: 98% CAS No.: 14315-97-0 Molecular Formula: (CH3O)2CHCH2CH2OCH3 Formula Weight: 134.17 Flash Point: 40°(105°F) Refractive Index: 1.4000 Boiling Point: 45-46°/17mm UN No.: 3271 MDL No.: MFCD00008490

Formule: C₆H₁₄O₃
Kookpunt: 45…46 °C (17 mmHg)
Dichtheid: 0,942
Vlampunt: 40 °C (105 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008490
CAS nummer: 14315-97-0
UN: 3271
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR