1-Decyl-3-methylimidazolium chloride ≥96.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1-decyl-3-methylimidazolium chloride

D5351-5G D5351-25G
TCIAD5351-5GEA 31.9 EUR
TCIAD5351-5G TCIAD5351-25G
1-Decyl-3-methylimidazolium chloride ≥96.0% (by HPLC)
1-Decyl-3-methylimidazolium chloride
Formule: C₁₄H₂₇ClN₂
MDL Nummer: MFCD03427605
CAS nummer: 171058-18-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=785ee8f186bf57c10516125307d5d162d6b38a0c&vid=785ee8f186bf57c10516125307d5d162d6b38a0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR