2-Decyltetradecan-1-amine ≥92.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D5359-1ML D5359-5ML
TCIAD5359-1MLEA 108 EUR
TCIAD5359-1ML TCIAD5359-5ML
2-Decyltetradecan-1-amine ≥92.0% (by GC, titration analysis)
2-Decyltetradecan-1-amine
Formule: C₂₄H₅₁N
MDL Nummer: MFCD26407439
CAS nummer: 62281-07-6
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 92.0 %
Purity (Neutralization titration) min. 92.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR