Diisopropyl Hydrazine-1,2-dicarboxylate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D5365-200MG D5365-1G
TCIAD5365-200MGEA 48.4 EUR
TCIAD5365-200MG TCIAD5365-1G
Diisopropyl Hydrazine-1,2-dicarboxylate ≥98.0% (by GC)
Diisopropyl Hydrazine-1,2-dicarboxylate
Formule: C₈H₁₆N₂O₄
Smeltpunt: 108°C
MDL Nummer: MFCD19443326
CAS nummer: 19740-72-8
EINECS: 243-263-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68dacc01d4be7b3cbd04ca546136de9f2089b5a9&vid=68dacc01d4be7b3cbd04ca546136de9f2089b5a9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Specificaties
Environmentally Preferable