2,5-Diphenylpyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D5456-1G D5456-5G
TCIAD5456-1GEA 113 EUR
TCIAD5456-1G TCIAD5456-5G
2,5-Diphenylpyridine ≥98.0% (by GC)
2,5-Diphenylpyridine
Formule: C₁₇H₁₃N
Kookpunt: 255°C
MDL Nummer: MFCD00444961
CAS nummer: 15827-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2daf915bfcbb7fab1f79712234c713c843f6cb6a&vid=2daf915bfcbb7fab1f79712234c713c843f6cb6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 171.0 to 175.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR