2-(2,4-Difluorophenyl)-5-methylpyridine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D5484-1G D5484-200MG
TCIAD5484-1GEA 118 EUR
TCIAD5484-1G TCIAD5484-200MG
2-(2,4-Difluorophenyl)-5-methylpyridine ≥98.0% (by GC, titration analysis)
2-(2,4-Difluorophenyl)-5-methylpyridine
Formule: C₁₂H₉F₂N
MDL Nummer: MFCD17170319
CAS nummer: 583052-21-5

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR