Di(N-succinimidyl) L-Tartrate

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D5500-50MG D5500-250MG
TCIAD5500-50MGEA 86.9 EUR
TCIAD5500-50MG TCIAD5500-250MG
Di(N-succinimidyl) L-Tartrate
Di(N-succinimidyl) L-Tartrate
Formule: C₁₂H₁₂N₂O₁₀
CAS nummer: 62069-75-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Elemental analysis (Nitrogen) 7.70 to 8.50 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR