2,6-Diethyl-4-methylaniline ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D5517-1G D5517-5G
TCIAD5517-1GEA 23.1 EUR
TCIAD5517-1G TCIAD5517-5G
2,6-Diethyl-4-methylaniline ≥98.0% (by GC)
2,6-Diethyl-4-methylaniline
Formule: C₁₁H₁₇N
MW: 163.26 g/mol
Kookpunt: 121°C
MDL Nummer: MFCD00035950
CAS nummer: 24544-08-9
EINECS: 246-307-5
Order Now
Specification Test Results
Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %