Dodecylamine Hydroiodide ≥98.0% (by total nitrogen and titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Dodecylammonium iodide, Dodecan-1-amine Hydroiodide, Laurylamine Hydroiodide, 1-Aminododecane Hydroiodide
D5538-1G
D5538-5G
TCIAD5538-1GEA
36.3
EUR
TCIAD5538-1G
TCIAD5538-5G
Dodecylamine Hydroiodide ≥98.0% (by total nitrogen and titration analysis)
Dodecylamine Hydroiodide
Formule:
C₁₂H₂₇N·HI MW: 313.27 g/mol |
CAS nummer:
34099-97-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...