2,5-Diiodo-p-xylene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D5589-1G D5589-5G
TCIAD5589-1GEA 70.4 EUR
TCIAD5589-1G TCIAD5589-5G
2,5-Diiodo-p-xylene ≥98.0% (by GC)
2,5-Diiodo-p-xylene
Formule: C₈H₈I₂
MDL Nummer: MFCD00829126
CAS nummer: 1124-08-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR