Diethyl phenylphosphonite ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D5593-1G D5593-5G
TCIAD5593-1GEA 24.2 EUR
TCIAD5593-1G TCIAD5593-5G
Diethyl phenylphosphonite ≥96.0% (by GC)
Diethyl phenylphosphonite
Formule: C₁₀H₁₅O₂P
Kookpunt: 235°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009086
CAS nummer: 1638-86-4
EINECS: 216-676-7
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR