O,O'-Dimethyl Dithiophosphate ≥85.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D5602-1G D5602-5G
TCIAD5602-1GEA 64.9 EUR
TCIAD5602-1G TCIAD5602-5G
O,O'-Dimethyl Dithiophosphate ≥85.0% (by titrimetric analysis)
O,O'-Dimethyl Dithiophosphate
Formule: C₂H₇O₂PS₂
Kookpunt: 57°C
Vlampunt: 48°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00127999
CAS nummer: 756-80-9
EINECS: 212-053-9
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Red clear liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR