N,N-Dimethylethylenediamine dihydroiodide ≥98.0% (by total nitrogen basis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
N,N-Dimethylethylenediammonium Diiodide, N1,N1-Dimethylethane-1,2-diamine dihydroiodide, 2-(Dimethylamino)ethylamine dihydroiodide
D5616-1G
D5616-5G
TCIAD5616-1GEA
27.5
EUR
TCIAD5616-1G
TCIAD5616-5G
N,N-Dimethylethylenediamine dihydroiodide ≥98.0% (by total nitrogen basis)
N,N-Dimethylethylenediamine dihydroiodide
Formule:
C₄H₁₂N₂·₂HI |
CAS nummer:
244234-52-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...