Dimethyl 2-(4-Methoxybenzylidene)malonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-(4-Methoxybenzylidene)malonic acid Dimethyl ester

D5642-25G D5642-5G
TCIAD5642-25GEA 112 EUR
TCIAD5642-25G TCIAD5642-5G
Dimethyl 2-(4-Methoxybenzylidene)malonate ≥98.0% (by GC)
Dimethyl 2-(4-Methoxybenzylidene)malonate
Formule: C₁₃H₁₄O₅
Kookpunt: 130°C
MDL Nummer: MFCD09753105
CAS nummer: 7443-25-6
EINECS: 231-185-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR