2-Diphenylphosphino-6-methylpyridine ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D5683-1G D5683-5G
TCIAD5683-1GEA 78.1 EUR
TCIAD5683-1G TCIAD5683-5G
2-Diphenylphosphino-6-methylpyridine ≥95.0% (by GC)
2-Diphenylphosphino-6-methylpyridine
Formule: C₁₈H₁₆NP
MDL Nummer: MFCD04974234
CAS nummer: 132682-77-0

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 93.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR