[Dimethoxy(methyl)silyl]methyl methacrylate ≥90.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

D5699-5G D5699-25G
TCIAD5699-5GEA 52.8 EUR
TCIAD5699-5G TCIAD5699-25G
[Dimethoxy(methyl)silyl]methyl methacrylate ≥90.0% (by GC) stabilised
[Dimethoxy(methyl)silyl]methyl methacrylate
Formule: C₈H₁₆O₄Si
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD07777082
CAS nummer: 121177-93-3

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR