N-Ethyl-1-naphthylamine hydrobromide ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E0149-25G E0149-5G
TCIAE0149-25GEA 131 EUR
TCIAE0149-25G TCIAE0149-5G
N-Ethyl-1-naphthylamine hydrobromide ≥98.0% (by titrimetric analysis)
N-Ethyl-1-naphthylamine hydrobromide
Formule: C₁₂H₁₃N
MW: 252.16 g/mol
MDL Nummer: MFCD00060154
CAS nummer: 36966-04-8
EINECS: 253-291-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR