N-Ethylcarbazole-3-carboxaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
E0359-25G
E0359-5G
TCIAE0359-25GEA
171
EUR
TCIAE0359-25G
TCIAE0359-5G
N-Ethylcarbazole-3-carboxaldehyde ≥98.0% (by GC)
N-Ethylcarbazole-3-carboxaldehyde
Formule:
C₁₅H₁₃NO Kookpunt: 255°C |
MDL Nummer:
MFCD00004963 CAS nummer: 7570-45-8 EINECS: 231-471-2 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 91.0 deg-C |
Solubility in Toluene | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...