4,6-O-Ethylidene-ɑ-D-glucopyranose ≥92.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
4,6-O-Ethylidene-ɑ-D-glucose, 4,6-O-Ethylidene-D-glucopyranose
E0402-1G
E0402-5G
TCIAE0402-1GEA
45.1
EUR
TCIAE0402-1G
TCIAE0402-5G
4,6-O-Ethylidene-ɑ-D-glucopyranose ≥92.0% (by GC)
4,6-O-Ethylidene-alpha-D-glucopyranose
Formule:
C₈H₁₄O₆ |
MDL Nummer:
MFCD00006820 CAS nummer: 13224-99-2 EINECS: 236-198-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 92.0 % |
Specific rotation [a]20/D | -2.0 to -4.5 deg (c=1,H2O) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...