Vinyl-2-ethylhexanoate ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

E0404-25ML E0404-500ML
TCIAE0404-25MLEA 23.1 EUR
TCIAE0404-25ML TCIAE0404-500ML
Vinyl-2-ethylhexanoate ≥98.0% (by GC) stabilised
Vinyl-2-ethylhexanoate
Formule: C₁₀H₁₈O₂
Kookpunt: 86°C
Vlampunt: 65°C
MDL Nummer: MFCD00009488
CAS nummer: 94-04-2
EINECS: 202-297-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR