Ethyl-2-ethyl-2-methylacetoacetate ≥93.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E0437-25ML E0437-5ML
TCIAE0437-25MLEA 128 EUR
TCIAE0437-25ML TCIAE0437-5ML
Ethyl-2-ethyl-2-methylacetoacetate ≥93.0% (by GC)
Ethyl-2-ethyl-2-methylacetoacetate
Formule: C₉H₁₆O₃
Kookpunt: 199°C
MDL Nummer: MFCD00191433
CAS nummer: 33697-53-9

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR