Ethyl glutaryl chloride ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Ethyl 5-chloro-5-oxopentanoate, Glutaric Acid Monoethyl Ester Chloride, Ethyl 4-(Chloroformyl)butyrate
E0451-25G
E0451-5G
TCIAE0451-25GEA
91.3
EUR
TCIAE0451-25G
TCIAE0451-5G
Ethyl glutaryl chloride ≥98.0% (by titrimetric analysis)
Ethyl glutaryl chloride
Formule:
C₇H₁₁ClO₃ MW: 178.61 g/mol Kookpunt: 108°C Vlampunt: 82°C |
MDL Nummer:
MFCD00013659 CAS nummer: 5205-39-0 EINECS: 226-000-2 UN: 3265 ADR: 8 III |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...