4-Ethoxycoumarin ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
4-Ethoxy-2H-chromen-2-one
E0545-25G
E0545-5G
TCIAE0545-25GEA
162
EUR
TCIAE0545-25G
TCIAE0545-5G
4-Ethoxycoumarin ≥98.0% (by GC)
4-Ethoxycoumarin
Formule:
C₁₁H₁₀O₃ |
MDL Nummer:
MFCD00052755 CAS nummer: 35817-27-7 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...