4-Ethynylanisole ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E0603-1G
E0603-5G
E0603-25G
TCIAE0603-1GEA
33
EUR
TCIAE0603-1G
TCIAE0603-5G
TCIAE0603-25G
4-Ethynylanisole ≥98.0% (by GC)
4-Ethynylanisole
Formule:
C₉H₈O Kookpunt: 86°C Vlampunt: 82°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00168815 CAS nummer: 768-60-5 |
Specification Test Results
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...