2-(2-Ethoxyethoxy)ethyl acrylate ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Di(ethylene glycol) ethyl ether acrylate, Acrylic Acid 2-(2-Ethoxyethoxy)ethyl Ester, Diethylene Glycol Monoethyl Ether Acrylate

Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

E0652-25G E0652-500G
TCIAE0652-25GEA 17.6 EUR
TCIAE0652-25G TCIAE0652-500G
2-(2-Ethoxyethoxy)ethyl acrylate ≥98.0% (by GC) stabilised
2-(2-Ethoxyethoxy)ethyl acrylate
Formule: C₉H₁₆O₄
MW: 188.22 g/mol
MDL Nummer: MFCD00015655
CAS nummer: 7328-17-8
EINECS: 230-811-7

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR