Ethyl-5-bromo-1H-indole-2-carboxylate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Ethyl 5-bromoindole-2-carboxylate, 5-Bromoindole-2-carboxylic acid ethyl ester, Ethyl 5-Bromoindole-2-carboxylate, Ethyl 5-bromo-1H-indole-2-carboxylate
E0698-1G
E0698-5G
TCIAE0698-1GEA
24.2
EUR
TCIAE0698-1G
TCIAE0698-5G
Ethyl-5-bromo-1H-indole-2-carboxylate ≥98.0% (by GC)
Ethyl-5-bromo-1H-indole-2-carboxylate
Formule:
C₁₁H₁₀BrNO₂ |
MDL Nummer:
MFCD00022701 CAS nummer: 16732-70-0 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 164.0 to 168.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...