N-Ethyl-2-isopropyl-5-methylcyclohexanecarboxamide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

E0796-25G E0796-5G
TCIAE0796-25GEA 83.6 EUR
TCIAE0796-25G TCIAE0796-5G
N-Ethyl-2-isopropyl-5-methylcyclohexanecarboxamide ≥98.0% (by GC)
N-Ethyl-2-isopropyl-5-methylcyclohexanecarboxamide
Formule: C₁₃H₂₅NO
MDL Nummer: MFCD00130071
CAS nummer: 39711-79-0
EINECS: 254-599-0
Merck Index: 14,03825

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR