Ethyl-3-methylvalerate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

E0803-25G E0803-5G
TCIAE0803-25GEA 141 EUR
TCIAE0803-25G TCIAE0803-5G
Ethyl-3-methylvalerate ≥98.0% (by GC)
Ethyl-3-methylvalerate
Formule: C₈H₁₆O₂
Kookpunt: 158°C
Vlampunt: 45°C
MDL Nummer: MFCD00041390
CAS nummer: 5870-68-8
EINECS: 227-524-4
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR