1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E0806-1G
E0806-5G
TCIAE0806-1GEA
31.9
EUR
TCIAE0806-1G
TCIAE0806-5G
1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene ≥98.0% (by GC)
1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene
Formule:
C₁₇H₁₆O |
MDL Nummer:
MFCD00807428 CAS nummer: 63221-88-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...