Ethanolamine thioglycolate ∼85% in water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Ethanolamine Mercaptoacetate, Monoethanolamine Thioglycolate

E0813-25G E0813-500G
TCIAE0813-25GEA 17.6 EUR
TCIAE0813-25G TCIAE0813-500G
Ethanolamine thioglycolate ∼85% in water
Ethanolamine thioglycolate
Formule: C₄H₁₁NO₃S
MDL Nummer: MFCD00050789
CAS nummer: 126-97-6
EINECS: 204-815-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (Iodometric titration) 83.0 to 88.0 w/w%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR