7-Ethyl-3-indoleethanol 98,0 GC_ASSAY_METHOD

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-(7-Ethyl-1H-indol-3-yl)ethanol, 7-Ethyltryptophol, 7-Ethyl-3-(2-hydroxyethyl)indole, Etodolac impurity H

E0824-25G E0824-5G
TCIAE0824-25GEA 338 EUR
TCIAE0824-25G TCIAE0824-5G
7-Ethyl-3-indoleethanol 98,0 GC_ASSAY_METHOD
7-Ethyl-3-indoleethanol
Formule: C₁₂H₁₅NO
MW: 189.26 g/mol
MDL Nummer: MFCD01718805
CAS nummer: 41340-36-7
EINECS: 431-020-1
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR